siRNA Details
| Gene Symbol: Dgke | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019505.3 | NP_062378.1 | 1 | 801 | ||
| GenBank mRNA | AF136744.1 | - | 1 | 438 | ||
| AK044596.1 | - | 1537 | ||||
| AK049335.1 | - | 1 | 601 | |||
| AK147841.1 | - | 1 | 802 | |||
| BB635763.1 | - | 1 | 205 | |||
| BC055867.1 | - | 1 | 734 | |||

Dgke
Genomic Map