siRNA Details
| Gene Symbol: Prpf40a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018785.2 | NP_061255.1 | 3, 4 | 380 | ||
| GenBank mRNA | AF135439.1 | - | 3, 4 | 380 | ||
| AK045207.1 | - | 3, 4 | 398 | |||
| AK087963.1 | - | 3, 4 | 297 | |||
| BC003318.1 | - | 3, 4 | 407 | |||

Prpf40a
Genomic Map