siRNA Details
| Gene Symbol: Uchl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159866.1 | NP_001153338.1 | 10 | 1035 | ||
| NM_019562.2 | NP_062508.2 | 10 | 1038 | |||
| GenBank mRNA | AF148447.1 | - | 10 | 981 | ||
| AF175903.1 | - | 10 | 987 | |||
| AK007860.1 | - | 9 | 849 | |||
| AK011117.1 | - | 10 | 1005 | |||
| AK017925.1 | - | 10 | 1033 | |||
| BC006891.1 | - | 10 | 988 | 2650799 | ||

Uchl5
Genomic Map