siRNA Details
Gene Symbol: Uchl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159866.1 | NP_001153338.1 | 5 | 517 | ||
NM_019562.2 | NP_062508.2 | 5 | 517 | |||
GenBank mRNA | AF148447.1 | - | 5 | 460 | ||
AF175903.1 | - | 5 | 469 | |||
AK007860.1 | - | 4 | 331 | |||
AK011117.1 | - | 5 | 484 | |||
AK017925.1 | - | 5 | 515 | |||
BC006891.1 | - | 5 | 470 | 2650799 |
