siRNA Details
| Gene Symbol: Htra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019564.3 | NP_062510.2 | 5 | 1055 | ||
| GenBank mRNA | AF172994.1 | - | 5 | 1031 | ||
| AF179369.1 | - | 5 | 1026 | |||
| AK090320.1 | - | 5 | 770 | |||
| AK090321.1 | - | 2 | 202 | |||
| AK128916.1 | - | 5 | 1028 | |||
| AK132270.1 | - | 5 | 1028 | |||
| AK167667.1 | - | 5 | 621 | |||
| BC013516.1 | - | 5 | 1025 | 4235912 | ||

Htra1
Genomic Map