siRNA Details
| Gene Symbol: Tspan5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019571.5 | NP_062517.1 | 7 | 1058 | ||
| GenBank mRNA | AF121344.1 | - | 7 | 1073 | ||
| AK013350.1 | - | 7 | 1023 | |||
| AK015705.1 | - | 7 | 780 | |||
| AK033543.1 | - | 7 | 757 | |||
| BC046784.1 | - | 7 | 897 | 6416107 | ||
| BC058695.1 | - | 7 | 1029 | 6813872 | ||

Tspan5
Genomic Map