siRNA Details
| Gene Symbol: Tspan5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019571.5 | NP_062517.1 | 7, 8 | 1101 | ||
| GenBank mRNA | AF121344.1 | - | 7, 8 | 1116 | ||
| AK013350.1 | - | 7, 8 | 1066 | |||
| AK015705.1 | - | 7, 8 | 823 | |||
| AK033543.1 | - | 7, 8 | 800 | |||
| BC046784.1 | - | 7, 8 | 940 | 6416107 | ||
| BC058695.1 | - | 7, 8 | 1072 | 6813872 | ||

Tspan5
Genomic Map