siRNA Details
| Gene Symbol: Ube2j1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019586.3 | NP_062532.2 | 2, 3 | 262 | ||
| GenBank mRNA | AJ245899.1 | - | 2, 3 | 196 | ||
| AK003050.1 | - | 2, 3 | 194 | |||
| AK031669.1 | - | 2, 3 | 150 | |||
| AK149985.1 | - | 2, 3 | 173 | |||
| AK160428.1 | - | 2, 3 | 268 | |||
| BC024623.1 | - | 2, 3 | 200 | 4217620 | ||
| BY707310.1 | - | 2, 3 | 262 | |||

Ube2j1
Genomic Map