siRNA Details
| Gene Symbol: Stk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019635.2 | NP_062609.2 | 6 | 919 | ||
| GenBank mRNA | AF271361.1 | - | 6 | 665 | ||
| AK002914.1 | - | 1 | 137 | |||
| AY058922.1 | - | 6 | 805 | |||
| BC037440.1 | - | 4 | 675 | 3585312 | ||
| BC049123.1 | - | 6 | 793 | 6813956 | ||
| U28726.1 | - | 6 | 798 | |||

Stk3
Genomic Map