siRNA Details
| Gene Symbol: Rbm14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019869.3 | NP_063922.2 | 2 | 1140 | ||
| GenBank mRNA | AK004928.1 | - | 2 | 1110 | ||
| AK050945.1 | - | 2 | 1142 | |||
| AK088201.1 | - | 2 | 1106 | |||
| AK153230.1 | - | 3 | 1016 | |||
| AK167503.1 | - | 2 | 1119 | |||
| BC010294.1 | - | 2 | 1091 | 3153900 | ||

Rbm14
Genomic Map