siRNA Details
Gene Symbol: Ggcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019802.5 | NP_062776.1 | 13, 14 | 1975 | ||
GenBank mRNA | AF087938.1 | - | 13, 14 | 1950 | ||
AK033496.1 | - | 13, 14 | 1902 | |||
AK036577.1 | - | 13, 14 | 1949 | |||
AK135425.1 | - | 13, 14 | 1962 | |||
AK165982.1 | - | 13, 14 | 1950 | |||
BC138763.1 | - | 13, 14 | 1900 | 8861785 | ||
BC138764.1 | - | 13, 14 | 1900 | 8861786 |
