siRNA Details
| Gene Symbol: Stam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019667.2 | NP_062641.1 | 7 | 637 | ||
| GenBank mRNA | AB012611.1 | - | 7 | 652 | ||
| AK004604.2 | - | 7 | 637 | |||
| AK035789.1 | - | 690 | ||||
| AK044230.1 | - | 6 | 566 | |||
| AK078244.1 | - | 7 | 669 | |||
| AK159699.1 | - | 7 | 666 | |||
| AK168729.1 | - | 7 | 669 | |||
| BC013818.1 | - | 7 | 673 | 3671426 | ||
| DQ141211.1 | - | 8 | 867 | |||

Stam2
Genomic Map