siRNA Details
| Gene Symbol: Abcb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019875.2 | NP_063928.2 | 10 | 2017 | ||
| GenBank mRNA | AB045382.2 | - | 9 | 1837 | ||
| AB093298.1 | - | 10 | 1999 | |||
| AF216495.1 | - | 10 | 2010 | |||
| AK020749.1 | - | 4 | 717 | |||
| AK044140.1 | - | 10 | 1995 | |||
| BC053014.1 | - | 10 | 1970 | 5686698 | ||

Abcb9
Genomic Map