siRNA Details
| Gene Symbol: Net1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047159.2 | NP_001040624.1 | 10 | 1477 | ||
| NM_019671.3 | NP_062645.2 | 12 | 1598 | |||
| GenBank mRNA | AF094520.1 | - | 12 | 1494 | ||
| AJ010045.1 | - | 10 | 1440 | |||
| AK082128.1 | - | 10 | 1474 | |||
| AK088632.1 | - | 10 | 1441 | |||
| AK139921.1 | - | 10 | 1441 | |||
| BC004699.1 | - | 10 | 1451 | 3499258 | ||
| BG083317.2 | - | 3 | 264 | |||

Net1
Genomic Map