siRNA Details
| Gene Symbol: Cpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019775.3 | NP_062749.2 | 11 | 1277 | ||
| GenBank mRNA | AB021968.1 | - | 11 | 1225 | ||
| AF164524.1 | - | 11 | 1292 | |||
| AF186188.1 | - | 11 | 1228 | |||
| AK004045.1 | - | 11 | 1250 | |||
| AK015095.1 | - | 894 | ||||
| BC089577.1 | - | 11 | 1249 | 6766672 | ||
| CO807338.1 | - | 3 | 311 | |||

Cpb2
Genomic Map