siRNA Details
| Gene Symbol: Cpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019775.3 | NP_062749.2 | 7 | 750 | ||
| GenBank mRNA | AB021968.1 | - | 7 | 698 | ||
| AF164524.1 | - | 7 | 765 | |||
| AF186188.1 | - | 7 | 701 | |||
| AK004045.1 | - | 7 | 723 | |||
| BC089577.1 | - | 7 | 722 | 6766672 | ||

Cpb2
Genomic Map