siRNA Details
| Gene Symbol: Kcnj1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168354.1 | NP_001161826.1 | 2 | 498 | ||
| NM_019659.3 | NP_062633.1 | 2 | 496 | |||
| GenBank mRNA | AF012834.1 | - | 2 | 317 | ||
| AK085262.1 | - | 2 | 422 | |||
| BC020525.1 | - | 2 | 498 | 4241262 | ||
| CF168846.1 | - | 2 | 389 | |||

Kcnj1
Genomic Map