siRNA Details
| Gene Symbol: Spen | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019763.2 | NP_062737.2 | 12 | 6301 | ||
| GenBank mRNA | AB055980.1 | - | 10 | 5674 | ||
| AF156529.1 | - | 11 | 5803 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spen| Gene Symbol: Spen | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019763.2 | NP_062737.2 | 12 | 6301 | ||
| GenBank mRNA | AB055980.1 | - | 10 | 5674 | ||
| AF156529.1 | - | 11 | 5803 | |||