siRNA Details
| Gene Symbol: Akap8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB028920.1 | - | 12 | 1529 | ||
| AK004801.1 | - | 12 | 1535 | |||
| AK089092.1 | - | 12 | 1542 | |||
| BC042602.1 | - | 12 | 2412 | |||
| BC054433.1 | - | 10 | 2109 | |||
| BC060170.1 | - | 12 | 2409 | |||
| BC060209.1 | - | 11 | 2025 | |||
| BC078448.1 | - | 11 | 1915 | |||
| BC125521.1 | - | 12 | 1525 | 40130036 | ||
| BC137746.1 | - | 12 | 1525 | 8860765 | ||

Akap8
Genomic Map