siRNA Details
| Gene Symbol: Adar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038587.4 | NP_001033676.2 | 6 | 2167 | ||
| NM_001146296.1 | NP_001139768.1 | 6 | 2159 | |||
| NM_019655.3 | NP_062629.3 | 6 | 2159 | |||
| GenBank mRNA | AF052506.1 | - | 6 | 2072 | ||
| AF291050.1 | - | 6 | 2066 | |||
| AF291875.1 | - | 5 | 634 | |||
| AF291876.1 | - | 6 | 2066 | |||
| AF291877.1 | - | 5 | 634 | |||
| AK076413.1 | - | 6 | 2118 | |||
| AK089451.1 | - | 6 | 2130 | |||
| AK147614.1 | - | 6 | 2130 | |||
| AK156093.1 | - | 5 | 726 | |||
| AK172523.1 | - | 5 | 726 | |||
| BC042505.1 | - | 6 | 2093 | 4012994 | ||
| CT010323.1 | - | 6 | 2066 | |||

Adar
Genomic Map