siRNA Details
| Gene Symbol: Diap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019670.1 | NP_062644.1 | 3 | 299 | ||
| GenBank mRNA | AF094519.1 | - | 3 | 299 | ||
| AK137073.1 | - | 2 | 388 | |||
| AK138694.1 | - | 3 | 329 | |||
| AK161737.1 | - | 2 | 317 | |||
| AK166225.1 | - | 3 | 329 | |||
| AK166882.1 | - | 3 | 355 | |||
| AK220493.1 | - | 3 | 363 | |||
| BC003250.1 | - | 2 | 333 | |||
| BC085191.1 | - | 3 | 409 | 30693873 | ||
| BC086779.1 | - | 3 | 374 | |||

Diap3
Genomic Map