siRNA Details
| Gene Symbol: Ppp4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019674.3 | NP_062648.1 | 8 | 834 | ||
| GenBank mRNA | AF088911.1 | - | 8 | 859 | ||
| AK027923.1 | - | 6 | 614 | |||
| AK144244.1 | - | 7 | 761 | |||
| AK199030.1 | - | 1 | 61 | |||
| BC001993.1 | - | 8 | 834 | 3491568 | ||
| U79747.1 | - | 4 | 486 | |||

Ppp4c
Genomic Map