siRNA Details
| Gene Symbol: Clip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291229.1 | NP_001278158.1 | 9 | 2004 | ||
| NM_019765.5 | NP_062739.2 | 9 | 2004 | |||
| GenBank mRNA | AK014540.1 | - | 9 | 1775 | ||
| AK029945.1 | - | 9 | 1841 | |||
| AK220172.1 | - | 9 | 1965 | |||
| BC007191.1 | - | 9 | 1944 | 3592614 | ||
| BC029243.1 | - | 6 | 1126 | |||
| BC052632.1 | - | 7 | 1410 | |||

Clip1
Genomic Map