siRNA Details
| Gene Symbol: Actl6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019673.2 | NP_062647.2 | 12 | 1401 | ||
| GenBank mRNA | AF041476.1 | - | 12 | 1221 | ||
| AK002724.1 | - | 12 | 1196 | |||
| AK090154.1 | - | 12 | 1220 | |||
| AK167905.1 | - | 12 | 1215 | |||
| AK169278.1 | - | 12 | 1220 | |||
| AK176960.1 | - | 2 | 111 | |||
| BC001994.1 | - | 12 | 1189 | 3491205 | ||
| BC094498.1 | - | 12 | 1217 | 4209491 | ||

Actl6a
Genomic Map