siRNA Details
| Gene Symbol: Actl6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019673.2 | NP_062647.2 | 2, 3 | 406 | ||
| GenBank mRNA | AF041476.1 | - | 2, 3 | 226 | ||
| AK002724.1 | - | 2, 3 | 202 | |||
| AK090154.1 | - | 2, 3 | 225 | |||
| AK167905.1 | - | 2, 3 | 220 | |||
| AK169278.1 | - | 2, 3 | 225 | |||
| AK182047.1 | - | 1, 2 | 76 | |||
| AK205745.1 | - | 1, 2 | 64 | |||
| BC001994.1 | - | 2, 3 | 194 | 3491205 | ||
| BC094498.1 | - | 2, 3 | 222 | 4209491 | ||
| CF725603.1 | - | 2, 3 | 406 | |||

Actl6a
Genomic Map