siRNA Details
| Gene Symbol: Clptm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019649.2 | NP_062623.2 | 8 | 1041 | ||
| GenBank mRNA | AK148080.1 | - | 8 | 1023 | ||
| AK152410.1 | - | 8 | 1031 | |||
| AK152809.1 | - | 8 | 1031 | |||
| AK165742.1 | - | 7 | 1735 | |||
| AK171901.1 | - | 8 | 1032 | |||
| AK191893.1 | - | 1 | 104 | |||
| AK205055.1 | - | 1 | 81 | |||
| BC022172.1 | - | 8 | 1032 | 4193279 | ||
| D67067.1 | - | 8 | 1028 | |||

Clptm1
Genomic Map