siRNA Details
Gene Symbol: Snd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019776.2 | NP_062750.2 | 19 | 2423 | ||
GenBank mRNA | AB021491.1 | - | 19 | 2380 | ||
AK088028.1 | - | 19 | 2382 | |||
AK133837.1 | - | 19 | 2429 | |||
AK159837.1 | - | 19 | 2388 | |||
AK161591.1 | - | 19 | 2377 | |||
AK162432.1 | - | 19 | 2364 | |||
AK181132.1 | - | 2 | 157 | |||
BC007126.1 | - | 19 | 2370 | 3592548 |
