siRNA Details
| Gene Symbol: Socs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019654.2 | NP_062628.2 | 3 | 1674 | ||
| GenBank mRNA | AF033187.1 | - | 1 | 1232 | ||
| AK172997.1 | - | 1 | 218 | |||
| AK195293.1 | - | 1 | 28 | |||
| BC053015.1 | - | 3 | 1674 | 5702321 | ||

Socs5
Genomic Map