siRNA Details
| Gene Symbol: Socs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019654.2 | NP_062628.2 | 3 | 848 | ||
| GenBank mRNA | AF033187.1 | - | 1 | 406 | ||
| AK135099.1 | - | 2 | 776 | |||
| BC053015.1 | - | 3 | 848 | 5702321 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Socs5| Gene Symbol: Socs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019654.2 | NP_062628.2 | 3 | 848 | ||
| GenBank mRNA | AF033187.1 | - | 1 | 406 | ||
| AK135099.1 | - | 2 | 776 | |||
| BC053015.1 | - | 3 | 848 | 5702321 | ||