siRNA Details
| Gene Symbol: Slc2a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019741.3 | NP_062715.2 | 9 | 1482 | ||
| GenBank mRNA | AF161071.1 | - | 7 | 843 | ||
| AK029720.1 | - | 9 | 1483 | |||
| BC023500.1 | - | 7 | 858 | 5043284 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc2a5| Gene Symbol: Slc2a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019741.3 | NP_062715.2 | 9 | 1482 | ||
| GenBank mRNA | AF161071.1 | - | 7 | 843 | ||
| AK029720.1 | - | 9 | 1483 | |||
| BC023500.1 | - | 7 | 858 | 5043284 | ||