siRNA Details
Gene Symbol: Zbtb20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285805.1 | NP_001272734.1 | 10 | 1699 | ||
NM_019778.2 | NP_062752.2 | 8 | 1646 | |||
NM_181058.1 | NP_851401.1 | 6 | 1424 | |||
GenBank mRNA | AB221632.1 | - | 2 | 1111 | ||
AF185576.1 | - | 6 | 1469 | |||
AF194030.1 | - | 4 | 1261 | |||
AK005028.2 | - | 6 | 1425 | |||
AK034574.1 | - | 1 | 298 | |||
AY028963.1 | - | 3 | 1228 | |||
BC031114.1 | - | 7 | 1532 | 4971586 | ||
BC056446.1 | - | 10 | 1671 | 5703319 |
