siRNA Details
| Gene Symbol: Rnf138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB025010.1 | - | 4, 5 | 685 | ||
| AB025011.1 | - | 4, 5 | 711 | |||
| AK004238.1 | - | 3, 4 | 487 | |||
| AK010486.1 | - | 3, 4 | 476 | |||
| AK136138.1 | - | 3, 4 | 488 | |||
| BC003712.1 | - | 3, 4 | 451 | 3593899 | ||

Rnf138
Genomic Map