siRNA Details
| Gene Symbol: Rnf138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB025010.1 | - | 3 | 560 | ||
| AB025011.1 | - | 3 | 560 | |||
| AK004238.1 | - | 2 | 362 | |||
| AK010486.1 | - | 2 | 351 | |||
| AK013419.1 | - | 2 | 222 | |||
| AK136138.1 | - | 2 | 363 | |||
| BC003712.1 | - | 2 | 326 | 3593899 | ||

Rnf138
Genomic Map