siRNA Details
| Gene Symbol: Pmfbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019938.3 | NP_064322.1 | 9 | 1312 | ||
| GenBank mRNA | AB029919.1 | - | 9 | 1312 | ||
| AK133053.1 | - | 8 | 1368 | |||
| BC138245.1 | - | 9 | 1264 | 8861264 | ||
| BC138246.1 | - | 9 | 1264 | 8861265 | ||

Pmfbp1
Genomic Map