siRNA Details
| Gene Symbol: Hspb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019960.2 | NP_064344.1 | 1 | 245 | ||
| GenBank mRNA | AF203375.1 | - | 1 | 228 | ||
| AK009633.1 | - | 1 | 246 | |||
| BC065388.1 | - | 1 | 228 | 851802 | ||
| BC092376.1 | - | 1 | 215 | 30609467 | ||

Hspb3
Genomic Map