siRNA Details
| Gene Symbol: Pex3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164195.1 | NP_001157667.1 | 5 | 677 | ||
| NM_019961.3 | NP_064345.1 | 5 | 677 | |||
| GenBank mRNA | AF152996.1 | - | 5 | 600 | ||
| AK005315.1 | - | 5 | 533 | |||
| AK005978.1 | - | 4 | 374 | |||
| AK012827.1 | - | 5 | 656 | |||
| AK075820.1 | - | 5 | 592 | |||
| AK146934.1 | - | 5 | 656 | |||
| BC033415.1 | - | 5 | 559 | 5008068 | ||
| BC037606.1 | - | 5 | 640 | 5361347 | ||

Pex3
Genomic Map