siRNA Details
| Gene Symbol: Ick | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163780.1 | NP_001157252.1 | 10 | 1722 | ||
| NM_019987.2 | NP_064371.2 | 10 | 1650 | |||
| GenBank mRNA | AF225918.1 | - | 10 | 1490 | ||
| AK052178.1 | - | 3 | 619 | |||
| AK087484.1 | - | 10 | 1574 | |||
| AK133537.1 | - | 10 | 1589 | |||
| AK154185.1 | - | 10 | 1723 | |||
| AK173068.1 | - | 5 | 879 | |||
| BC028863.1 | - | 9 | 1407 | 4224269 | ||

Ick
Genomic Map