siRNA Details
| Gene Symbol: Clec7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020008.2 | NP_064392.2 | 5 | 653 | ||
| GenBank mRNA | AF262985.1 | - | 5 | 649 | ||
| AY534909.1 | - | 5 | 561 | |||
| BC027742.1 | - | 5 | 652 | 5036647 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec7a| Gene Symbol: Clec7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020008.2 | NP_064392.2 | 5 | 653 | ||
| GenBank mRNA | AF262985.1 | - | 5 | 649 | ||
| AY534909.1 | - | 5 | 561 | |||
| BC027742.1 | - | 5 | 652 | 5036647 | ||