siRNA Details
Gene Symbol: Crtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019922.2 | NP_064306.2 | 4 | 917 | ||
GenBank mRNA | AJ006469.1 | - | 4 | 875 | ||
AK017797.1 | - | 4 | 880 | |||
AK047506.1 | - | 4 | 873 | |||
BC049890.1 | - | 4 | 854 | 6306169 |
