siRNA Details
| Gene Symbol: Pigo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB038560.1 | - | 6 | 2756 | ||
| AK050819.1 | - | 7 | 2097 | |||
| AK220315.1 | - | 6 | 2704 | |||
| BC025857.1 | - | 1 | 237 | |||
| BC036977.1 | - | 7 | 2101 | |||
| BC092394.1 | - | 6 | 2774 | 30534262 | ||
| BC141058.1 | - | 6 | 2712 | 9055670 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_233141, XM_006238127.

Pigo
Genomic Map