siRNA Details
| Gene Symbol: Brinp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019967.2 | NP_064351.2 | 8 | 2477 | ||
| GenBank mRNA | AB060589.1 | - | 8 | 2477 | ||
| AF202896.1 | - | 7 | 2046 | |||
| AK028321.1 | - | 8 | 2392 | |||
| AK045420.1 | - | 8 | 2359 | |||
| BC079630.1 | - | 8 | 2444 | 5694980 | ||

Brinp1
Genomic Map