siRNA Details
| Gene Symbol: Alg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019998.3 | NP_064382.3 | 2 | 472 | ||
| GenBank mRNA | AB041604.1 | - | 2 | 409 | ||
| AB161357.1 | - | 2 | 406 | |||
| AK004997.2 | - | 2 | 430 | |||
| AK010673.1 | - | 2 | 438 | |||
| AK188072.1 | - | 2 | 62 | |||
| AK204608.1 | - | 2 | 62 | |||
| BC051951.1 | - | 2 | 420 | 5686826 | ||
| BC052411.1 | - | 2 | 425 | 5720077 | ||

Alg2
Genomic Map