siRNA Details
| Gene Symbol: Rec8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020002.3 | NP_064386.2 | 3, 4 | 375 | ||
| GenBank mRNA | AF262055.1 | - | 3, 4 | 305 | ||
| AK077167.1 | - | 3, 4 | 310 | |||
| AK146832.1 | - | 3, 4 | 361 | |||
| AU080225.1 | - | 3, 4 | 376 | |||
| BC052155.1 | - | 3, 4 | 371 | 6335959 | ||

Rec8
Genomic Map