siRNA Details
| Gene Symbol: Mbnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253708.1 | NP_001240637.1 | 4 | 1571 | ||
| NM_001253709.1 | NP_001240638.1 | 3 | 522 | |||
| NM_001253710.1 | NP_001240639.1 | 3 | 522 | |||
| NM_001253711.1 | NP_001240640.1 | 3 | 522 | |||
| NM_001253713.1 | NP_001240642.1 | 4 | 525 | |||
| NM_020007.3 | NP_064391.2 | 4 | 1571 | |||
| GenBank mRNA | AB093240.1 | - | 4 | 1575 | ||
| AF231110.1 | - | 3 | 1011 | |||
| AK038242.1 | - | 3 | 606 | |||
| AK153827.1 | - | 3 | 415 | |||
| AK153848.1 | - | 3 | 510 | |||
| BC060031.1 | - | 4 | 1549 | |||
| BC096600.1 | - | 3 | 483 | 4976244 | ||
| CN663545.1 | - | 1 | 98 | |||

Mbnl1
Genomic Map