siRNA Details
| Gene Symbol: Scube2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020052.2 | NP_064436.2 | 15 | 1754 | ||
| GenBank mRNA | AK016571.1 | - | 9 | 901 | ||
| AK142950.1 | - | 15 | 1720 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scube2| Gene Symbol: Scube2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020052.2 | NP_064436.2 | 15 | 1754 | ||
| GenBank mRNA | AK016571.1 | - | 9 | 901 | ||
| AK142950.1 | - | 15 | 1720 | |||