siRNA Details
| Gene Symbol: Hacl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019975.3 | NP_064359.2 | 4, 5 | 415 | ||
| GenBank mRNA | AJ132139.1 | - | 4, 5 | 413 | ||
| AK005505.1 | - | 4, 5 | 402 | |||
| AK041686.1 | - | 4, 5 | 411 | |||
| AK050078.1 | - | 4, 5 | 415 | |||
| AK161488.1 | - | 395 | ||||
| BC021360.1 | - | 4, 5 | 346 | 5011451 | ||

Hacl1
Genomic Map