siRNA Details
| Gene Symbol: Aldh1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053080.3 | NP_444310.3 | 10 | 1232 | ||
| GenBank mRNA | AF152359.1 | - | 10 | 1187 | ||
| AF246711.1 | - | 10 | 1180 | |||
| AF253409.1 | - | 10 | 1219 | |||
| AF280404.1 | - | 10 | 1184 | |||
| AK147006.1 | - | 10 | 1233 | |||
| BC038646.1 | - | 1 | 53 | |||
| BC048941.1 | - | 2 | 145 | |||
| BC058277.1 | - | 10 | 1176 | 6515355 | ||

Aldh1a3
Genomic Map