siRNA Details
| Gene Symbol: Rnf32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289757.1 | NP_001276686.1 | 3, 4 | 292 | ||
| NM_021470.6 | NP_067445.3 | 3, 4 | 264 | |||
| GenBank mRNA | AF190664.1 | - | 1 | 26 | ||
| AK005795.1 | - | 3, 4 | 219 | |||
| AK006379.1 | - | 3, 4 | 208 | |||
| AK012288.1 | - | 3, 4 | 292 | |||
| AK016026.1 | - | 3, 4 | 235 | |||
| BC050964.1 | - | 3, 4 | 207 | |||
| BC061103.1 | - | 3, 4 | 230 | |||
| BC089493.1 | - | 3, 4 | 207 | 6741898 | ||

Rnf32
Genomic Map