siRNA Details
| Gene Symbol: Gabrq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290435.1 | NP_001277364.1 | 7 | 1046 | ||
| NM_020488.2 | NP_065234.1 | 7 | 1046 | |||
| GenBank mRNA | AF189260.1 | - | 7 | 788 | ||
| AK038859.1 | - | 7 | 957 | |||
| AK156674.1 | - | 7 | 1047 | |||
| BC119074.1 | - | 7 | 786 | 8733823 | ||
| BC119076.1 | - | 7 | 786 | 8733825 | ||

Gabrq
Genomic Map