siRNA Details
| Gene Symbol: Tas2r105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020501.1 | NP_065247.1 | 1 | 466 | ||
| GenBank mRNA | BC125515.1 | - | 1 | 498 | 40130030 | |
| BC125541.1 | - | 1 | 498 | 40130056 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tas2r105| Gene Symbol: Tas2r105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020501.1 | NP_065247.1 | 1 | 466 | ||
| GenBank mRNA | BC125515.1 | - | 1 | 498 | 40130030 | |
| BC125541.1 | - | 1 | 498 | 40130056 | ||